Learn More
Instech Replacement universal swivel-to-tether clamp for Instech swivels. Adjusts to tethers of range of sizes.
SDP
Catalog No. 5081020
Encompass
The Instech CLAMP is a universal swivel-to-tether clamp designed to secure spring tethers to various swivel models. This essential accessory provides a reliable mechanical connection between the swivel and the protective tether, ensuring that the fluid line remains protected even during animal movement. It is compatible with a wide range of Instech spring tethers and stainless steel swivels, making it a versatile component for rodent infusion setups.  
Features
  • Application: Secures spring tethers to dual or single channel swivels
  • Compatibility: Mates with all Instech spring tethers and VAB™ systems
  • Design: Universal fit for 20ga, 22ga, and 25ga stainless steel swivels
  • Function: Prevents tether detachment during high-activity research
  • Material: Durable construction for repeated use and autoclaving
  • Packaging: Available in convenient packs of five
Catalog No. 50-810-20 Supplier Instech Supplier No. CLAMP
The Instech CLAMP is a universal swivel-to-tether clamp designed to secure spring tethers to various swivel models. This essential accessory provides a reliable mechanical connection between the swivel and the protective tether, ensuring that the fluid line remains protected even during animal movement. It is compatible with a wide range of Instech spring tethers and stainless steel swivels, making it a versatile component for rodent infusion setups.  
Features
  • Application: Secures spring tethers to dual or single channel swivels
  • Compatibility: Mates with all Instech spring tethers and VAB™ systems
  • Design: Universal fit for 20ga, 22ga, and 25ga stainless steel swivels
  • Function: Prevents tether detachment during high-activity research
  • Material: Durable construction for repeated use and autoclaving
  • Packaging: Available in convenient packs of five