Learn More
Instech Swivel clamp for MINF tethers
SDP
Catalog No. 5081143
Encompass
The Instech MCLAMP is a replacement swivel clamp specifically designed for use with the MINF mouse head block tether system. It allows for the direct attachment of the fine wire tether to the outlet tube of low-torque swivels, ensuring efficient torque transmission as the animal moves.
Features
  • Type: Slotted swivel clamp
  • Application: Connects the MINF wire tether to a fluid swivel
  • Compatibility: Designed for use with Instech low-torque swivels, such as models 375/25 or 375/D/22QM
  • Function: Facilitates the use of lightweight .010" diameter looped wire tethers instead of traditional spring tethers
  • Packaging: Typically sold in packages of 5
Catalog No. 50-811-43 Supplier Instech Supplier No. MCLAMP
The Instech MCLAMP is a replacement swivel clamp specifically designed for use with the MINF mouse head block tether system. It allows for the direct attachment of the fine wire tether to the outlet tube of low-torque swivels, ensuring efficient torque transmission as the animal moves.
Features
  • Type: Slotted swivel clamp
  • Application: Connects the MINF wire tether to a fluid swivel
  • Compatibility: Designed for use with Instech low-torque swivels, such as models 375/25 or 375/D/22QM
  • Function: Facilitates the use of lightweight .010" diameter looped wire tethers instead of traditional spring tethers
  • Packaging: Typically sold in packages of 5