Learn More
IDEX Health & Science Upchurch Swivel Barb Adapter | Polypropylene | 1/16in ID Tubing
Catalog No. NC1763494
Encompass_Preferred
These swivel barbed adapters have been engineered for use with soft-walled tubing and are uniquely designed, made of two captive pieces acting as a one-piece fitting. These 1/4-28 flat-bottom barbed connections spin freely from the nut to prevent twist during installation. Manufactured from polypropylene, these are ideal for use with most aqueous solutions. Options exist for several different tubing inner diameters.
Catalog No. NC1763494 Supplier IDEX Health & Science Upchurch Supplier No. D646
These swivel barbed adapters have been engineered for use with soft-walled tubing and are uniquely designed, made of two captive pieces acting as a one-piece fitting. These 1/4-28 flat-bottom barbed connections spin freely from the nut to prevent twist during installation. Manufactured from polypropylene, these are ideal for use with most aqueous solutions. Options exist for several different tubing inner diameters.