Learn More
TARGETMOL CHEMICALS INC SX-682 10MG
SDP
Catalog No. 502576240
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SX-682 is a potent selective and orally bioavailable inhibitor of CXCR1/2 has the potential to treat castration-resistant prostate cancer. purity: 99%
Catalog No. 50-257-6240 Supplier TARGETMOL CHEMICALS INC Supplier No. T849710MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SX-682 is a potent selective and orally bioavailable inhibitor of CXCR1/2 has the potential to treat castration-resistant prostate cancer. purity: 99%