Learn More
Medchemexpress LLC SX-682 | 1648843-04-2 | 99.7% | 467.20 | 100 MG
SDP
Catalog No. 5000364824
Encompass_Preferred
SX-682 is an orally bioavailable, potent allosteric inhibitor of CXCR1 and CXCR2. It can block tumor myeloid-derived suppressor cells (MDSCs) recruitment and enhance T cell activation and antitumor immunity.
  • Orally bioavailable
  • Potent allosteric inhibitor of CXCR1 and CXCR2
  • Blocks tumor myeloid-derived suppressor cells (MDSCs) recruitment
  • Enhances T cell activation and antitumor immunity
Catalog No. 50-003-64824 Supplier Medchemexpress LLC Supplier No. 100MGHY119339
SX-682 is an orally bioavailable, potent allosteric inhibitor of CXCR1 and CXCR2. It can block tumor myeloid-derived suppressor cells (MDSCs) recruitment and enhance T cell activation and antitumor immunity.
  • Orally bioavailable
  • Potent allosteric inhibitor of CXCR1 and CXCR2
  • Blocks tumor myeloid-derived suppressor cells (MDSCs) recruitment
  • Enhances T cell activation and antitumor immunity