Learn More
Selleck Chemical LLC SX-682-5mg
SDP
Catalog No. 502389733
Encompass_Preferred
SX-682 is an orally bioavailable small-molecule allosteric inhibitor of CXCR1 and CXCR2 that blocks tumor MDSC recruitment and enhances T cell activation and antitumor immunity.
Catalog No. 50-238-9733 Supplier Selleck Chemical LLC Supplier No. S89475MG
SX-682 is an orally bioavailable small-molecule allosteric inhibitor of CXCR1 and CXCR2 that blocks tumor MDSC recruitment and enhances T cell activation and antitumor immunity.