Learn More
Medchemexpress LLC SX-682 SMALL MOLECULE CXCR 5MG
SDP
Catalog No. NC1790637
Encompass_Preferred
1648843-04-2, MFCD28502254,Formula C₁₉H₁₄BF₄N₃O₄S, Purity: 98.52%, Molecular Weight 467.20, SX-682 is an orally bioavailable, potent allosteric inhibitor of CXCR1 and CXCR2. SX-682 can block tumor myeloid-derived suppressor cells (MDSCs) recruitment and enhance T cell activation and antitumor immunity.
Catalog No. NC1790637 Supplier Medchemexpress LLC Supplier No. HY1193395MG
1648843-04-2, MFCD28502254,Formula C₁₉H₁₄BF₄N₃O₄S, Purity: 98.52%, Molecular Weight 467.20, SX-682 is an orally bioavailable, potent allosteric inhibitor of CXCR1 and CXCR2. SX-682 can block tumor myeloid-derived suppressor cells (MDSCs) recruitment and enhance T cell activation and antitumor immunity.