Learn More
Medchemexpress LLC HY-128587 5mg Medchemexpress, SY-1365 CAS:1816989-16-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1285875MG
Encompass_Preferred
Medchemexpress, HY-128587 5mg SY-1365 CAS:1816989-16-8 SY-1365 is a potent and first-in-class selective CDK7 inhibitor, with a Ki of 17.4 nM. SY-1365 exhibits anti-proliferative and apoptotic effects in solid tumor cell lines. SY-1365 possesses anti-tumor activity in hematological and multiple aggressive solid tumors[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0114