Learn More
TARGETMOL CHEMICALS INC SY-5609 5MG
SDP
Catalog No. 502580199
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SY-5609 is a selective and noncovalent inhibitor of CDK7(KD = 0.065 nM) with antitumor activity and inhibits apoptosis. purity: 99%
Catalog No. 50-258-0199 Supplier TARGETMOL CHEMICALS INC Supplier No. T360385MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SY-5609 is a selective and noncovalent inhibitor of CDK7(KD = 0.065 nM) with antitumor activity and inhibits apoptosis. purity: 99%