Learn More
Medchemexpress LLC HY-138293 5mg , SY-5609 CAS:2417302-07-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1382935MG
Encompass_Preferred
HY-138293 5mg SY-5609 CAS: 2417302-07-7 SY-5609 (CDK7-IN-3) is an orally active, highly selective, noncovalent CDK7 inhibitor with a KD of 0.065 nM. SY-5609 shows poor inhibition on CDK2 (Ki=2600 nM), CDK9 (Ki=960 nM), CDK12 (Ki=870 nM). SY-5609 induces apoptosis in tumor cells and has antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9101