Learn More
Medchemexpress LLC SY-LB-35 | 2603461-70-5 | C15H11N3O | 1 ML
SDP
Catalog No. 5000374848
Encompass_Preferred
SY-LB-35 is a potent bone morphogenetic protein (BMP) receptor agonist that can significantly increase cell number and viability in the C2C12 myoblast cell line. It also shifts the cell cycle towards the S and G2/M phases.
  • Stimulates canonical Smad and non-canonical PI3K/Akt, ERK, p38, and JNK intracellular signaling pathways.
  • Increases cell number and cell viability in C2C12 myoblast cells.
  • Causes cell cycle shifting to S and G2/M phases in C2C12 cells.
Catalog No. 50-003-74848 Supplier Medchemexpress LLC Supplier No. HY15079510MM/1ML
SY-LB-35 is a potent bone morphogenetic protein (BMP) receptor agonist that can significantly increase cell number and viability in the C2C12 myoblast cell line. It also shifts the cell cycle towards the S and G2/M phases.
  • Stimulates canonical Smad and non-canonical PI3K/Akt, ERK, p38, and JNK intracellular signaling pathways.
  • Increases cell number and cell viability in C2C12 myoblast cells.
  • Causes cell cycle shifting to S and G2/M phases in C2C12 cells.