Learn More
MilliporeSigma Soybean Casein Digest Broth (TSB) in bottle with screw cap and septum, double packed, 12/pk
Catalog No. NC0550114
Encompass
The Soybean-Casein-Digest Broth is used for sterility testing. It is suitable for culturing of fungi and aerobic bacteria. The fluid is double packed for increasing the test method reliability.
Catalog No. NC0550114 Supplier MilliporeSigma Supplier No. STBMTSB12DP
The Soybean-Casein-Digest Broth is used for sterility testing. It is suitable for culturing of fungi and aerobic bacteria. The fluid is double packed for increasing the test method reliability.