Learn More
ABclonal Technology 2X Universal SYBR Green Fast qPCR Mix, 25 mL, SYBR Green I, ABclonal Hot Start Taq DNA Polymerase, Universal ROX Dye
SDP
Catalog No. NC2332350
Encompass_Preferred
The ABclonal 2X Universal SYBR Green Fast qPCR Mix is a highly efficient solution tailored for sensitive real-time quantitative PCR applications. This ready-to-use mix combines advanced components including a hot start polymerase, optimized buffers, and a proprietary dye, ensuring accurate results across a variety of qPCR instruments. Its formulation minimizes non-specific amplification and enhances specificity across diverse samples.
  • 2X concentrated format
  • Compatible with all qPCR instruments
  • Hot start capability for improved specificity
  • Includes SYBR Green I and universal ROX dye
  • Demonstrates high reaction efficiency (99%)
  • Features excellent freeze-thaw stability
  • Recommended for target lengths of 70-200 bp
  • Storage temperature of -20℃
  • Used for research purposes only
  • Protect from light during storage
Catalog No. NC2332350 Supplier ABclonal Technology Supplier No. RK21203
The ABclonal 2X Universal SYBR Green Fast qPCR Mix is a highly efficient solution tailored for sensitive real-time quantitative PCR applications. This ready-to-use mix combines advanced components including a hot start polymerase, optimized buffers, and a proprietary dye, ensuring accurate results across a variety of qPCR instruments. Its formulation minimizes non-specific amplification and enhances specificity across diverse samples.
  • 2X concentrated format
  • Compatible with all qPCR instruments
  • Hot start capability for improved specificity
  • Includes SYBR Green I and universal ROX dye
  • Demonstrates high reaction efficiency (99%)
  • Features excellent freeze-thaw stability
  • Recommended for target lengths of 70-200 bp
  • Storage temperature of -20℃
  • Used for research purposes only
  • Protect from light during storage