Learn More
Abcam SYBR Green I, 5MG
Catalog No. 502654867
Encompass_Preferred
MW 509.7 Da, Purity >=98%. SYBR Green I is a highly sensitive fluorescent DNA binding stain for staining of double stranded DNA (dsDNA) or oligonucleotides in agarose or polyacrylamide gels.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-4867 Supplier Abcam Supplier No. AB2854245MG
MW 509.7 Da, Purity >=98%. SYBR Green I is a highly sensitive fluorescent DNA binding stain for staining of double stranded DNA (dsDNA) or oligonucleotides in agarose or polyacrylamide gels.
The product is subject to the following: Abcam Restricted Use Statement