Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SYBR™ Green I Nucleic Acid Gel Stain, 10,000X concentrate in DMSO

Catalog No. S7563
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 mL
20 x 50 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S7563 500 μL
S7585 20 x 50 μL
S7567 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S7563 Supplier Invitrogen™ Supplier No. S7563
Only null left
Add to Cart
Edge
Add to Cart

Extremely sensitive stain for detecting double-stranded DNA (dsDNA) in agarose and polyacrylamide gels

SYBR Green I offers greater sensitivity for dsDNA, making it especially useful for assays where the presence of contaminating RNA or ssDNA might obscure results. With exceptionally low background fluorescence and spectral characteristics that closely match light sources and filter sets in existing instruments, SYBR Green I stain is ideal for use with laser scanners.
Product use 1mL stains 100 minigels.

  • Sensitivity: at least four times greater than ethidium bromide for DNA in agarose gels
  • Ease of use: gels soaked in diluted stain can be visualized without desalting
  • Compatibility: with UV transilluminators, gel documentation systems, and laser scanners
  • Flexibility: for use with a broad range of applications, including DNA typing, PCR-based assays, DNA damage assays, analysis of complex samples, and real-time PCR detection

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Supplied as a 10,000X concentrate in DMSO

Store at -20°C, protected from light in a dessicator.
Detection Location In-Gel Detection
Detection Method Fluorescence
Label or Dye SYBR Green I
Quantity 500 μL
Shipping Condition Room Temperature
Target Molecule DNA
Product Type Nucleic Acid Gel Stain

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.