Learn More
Roche Diagnostics KAPA SYBR FAST qPCR Kit | 100x20uL Reactions (Universal)
Catalog No. 501965281 Shop All Roche Diagnostics Products
Encompass
Kits include KAPA SYBR FAST qPCR Master Mix (2X), which contains KAPA SYBR FAST DNA Polymerase, reaction buffer, dNTPs, SYBR Green I dye, and MgCl2at a final concentration of 2.5 mM. *This item is non returnable.
This item is not returnable. View return policy
Catalog No. 50-196-5281 Supplier Roche Diagnostics Supplier No. 07959362001
Kits include KAPA SYBR FAST qPCR Master Mix (2X), which contains KAPA SYBR FAST DNA Polymerase, reaction buffer, dNTPs, SYBR Green I dye, and MgCl2at a final concentration of 2.5 mM. *This item is non returnable.