Learn More
Medchemexpress LLC HY-K0501 1 mL (100 rxns) Medchemexpress, SYBR Green qPCR Master Mix CAS: Purity:>98%
SDP
Catalog No. 501933323
Encompass_Preferred
Medchemexpress, HY-K0501 1 mL (100 rxns) SYBR Green qPCR Master Mix CAS: MCE SYBR Green qPCR Master Mix is provided as a simple-to-use, stabilized 2X formulation that includes all components for qPCR except sample DNA, primers and water. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3323 Supplier Medchemexpress LLC Supplier No. HYK05011 ML (100 RXNS)
Medchemexpress, HY-K0501 1 mL (100 rxns) SYBR Green qPCR Master Mix CAS: MCE SYBR Green qPCR Master Mix is provided as a simple-to-use, stabilized 2X formulation that includes all components for qPCR except sample DNA, primers and water. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.