Learn More
Takara Bio TB Green™ Premix Ex Taq™ (Tli RNase H Plus), 200 Rxns
SDP
Catalog No. 50444019
Encompass
TB Green Premix Ex Taq (Tli RNase H Plus) provides reduced PCR inhibition caused by the presence of double-stranded RNA/cDNA hybrids remaining after cDNA synthesis. This inhibition is common when using low or RNase H minus RTs, and with GC-rich templates and/or genes with poor expression. The presence of dsRNA/cDNA hybrids is a frequent cause of poor qPCR amplification and/or reaction failure. TB Green Premix Ex Taq (Tli RNase H Plus) prevents this potential problem at no additional cost and without requiring a separate RNase H digestion step prior to qPCR. 200 Rxns
Catalog No. 50-444-019 Supplier Takara Bio Supplier No. RR420A
TB Green Premix Ex Taq (Tli RNase H Plus) provides reduced PCR inhibition caused by the presence of double-stranded RNA/cDNA hybrids remaining after cDNA synthesis. This inhibition is common when using low or RNase H minus RTs, and with GC-rich templates and/or genes with poor expression. The presence of dsRNA/cDNA hybrids is a frequent cause of poor qPCR amplification and/or reaction failure. TB Green Premix Ex Taq (Tli RNase H Plus) prevents this potential problem at no additional cost and without requiring a separate RNase H digestion step prior to qPCR. 200 Rxns