Learn More
Sigma Aldrich Fine Chemicals Biosciences SYBRR Green Extract-N-AmpT
Catalog No. 502900670
Encompass
The SYBR Green Extract-N-Amp PCR ReadyMix is the specially formulated PCR enzyme blend contained in the SYBR Green Extract-N-Amp Plant PCR kit and SYBR Green Extract-N-Amp Tissue PCR Kit. It is intended to supplement these kits for researchers who perform more than one amplification per extracted sample.
Catalog No. 50-290-0670 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S43201.2ML
The SYBR Green Extract-N-Amp PCR ReadyMix is the specially formulated PCR enzyme blend contained in the SYBR Green Extract-N-Amp Plant PCR kit and SYBR Green Extract-N-Amp Tissue PCR Kit. It is intended to supplement these kits for researchers who perform more than one amplification per extracted sample.