Learn More
Sigma Aldrich Fine Chemicals Biosciences SYBRR Green JumpStartTM Ta
Catalog No. 502914346
Encompass
SYBR(R) Green I is ideal for quantifying any DNA sequence. The dye binds to double-stranded DNA and detection is monitored by measuring the increase in fluorescence throughout cycling.
Catalog No. 50-291-4346 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S4438500RXN
SYBR(R) Green I is ideal for quantifying any DNA sequence. The dye binds to double-stranded DNA and detection is monitored by measuring the increase in fluorescence throughout cycling.