Learn More
Sigma Aldrich Fine Chemicals Biosciences SYBRR Green JumpStartTM Ta
Supplier:  Sigma Aldrich Fine Chemicals Biosciences S4438100RXN
Encompass
SYBR(R) Green I is ideal for quantifying any DNA sequence. The dye binds to double-stranded DNA and detection is monitored by measuring the increase in fluorescence throughout cycling.
Catalog No. 50-290-4556