Learn More
INTACT GENOMICS INC ig SYBR Green qPCR 2X Master Mix 200 Reactions
Supplier:  INTACT GENOMICS INC 3354
Encompass
Intact Genomics SYBR Green qPCR 2x master mix is a ready-to-use cocktail containing all components except primers and template, for the amplification and detection of DNA by qPCR. The Ig SYBR Green qPCR 2x master mix with integrated chemically modified hot start Taq DNA polymerase, SYBR Green I fluorescent dye, ROX dye*, MgCl2, dNTPs and stabilizers. This master mix is ideal for high-throughput real-time PCR screening and validation. This features a high-quality hot start Taq DNA Polymerase for higher fidelity and better amplification.Applications Gene expression validation Absolute quantification Mutation detection Pathogen detection GMO characterization Genetic profilingBenefits Enhanced efficiency, specificity, and sensitivity Compatible with all real-time PCR instruments Superior gene expression results under various cycling conditions Robust and active for cDNA synthesis at temperatures up to 55 °C.
Catalog No. 50-000-83087