Learn More
Aviva Systems Biology Corp SYCP3 Antibody - N-terminal region (ARP58334_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58334_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SYCP3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SYCP3. Alternate names: COR1, MGC71888, SCP3, SPGF4ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-62285