Learn More
Aviva Systems Biology Corp SYDE1 Antibody - N-terminal region (ARP42682_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42682_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SYDE1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SYDE1. Alternate names: 7h3, FLJ13511ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Rabbit, Rat
Catalog No. 50-014-58304