Learn More
Quality Biological Inc Syringe Filter, PES, 0.22um, 13mm, 75/CS
SDP
Supplier:  Quality Biological Inc P886229746
Encompass_Preferred
Syringe Filter, PES, 0.22um, 13mm, 75/CS
Catalog No. 50-751-7915