Learn More
Medchemexpress LLC HY-131341 5mg Medchemexpress, Syk-IN-4 CAS: Purity:>98%
SDP
Catalog No. 502081275
Encompass_Preferred
Medchemexpress, HY-131341 5mg Syk-IN-4 CAS: Syk-IN-4 is a potent, selective and orally bioavailable SYK inhibitor with an IC50 of 0.31 nM. SYK has emerged as a potential target for autoimmunity and hematological cancers. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1275 Supplier Medchemexpress LLC Supplier No. HY1313415MG
Medchemexpress, HY-131341 5mg Syk-IN-4 CAS: Syk-IN-4 is a potent, selective and orally bioavailable SYK inhibitor with an IC50 of 0.31 nM. SYK has emerged as a potential target for autoimmunity and hematological cancers. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.