Learn More
Cayman Chemical Syk Inhibitor, Purity: ≥95% (mixture of isomers), CAS: 622387-85-3, 1 mg
SDP
Catalog No. 502973015
Encompass_Preferred
An oxindole compound that potently blocks Syk activity (IC50 = 14 nM) and inhibits FcεRI-mediated rat RBL-2H3 basophil cell degranulation (EC50 = 313 nM); 2 μM completely abolishes convulxin-induced platelet aggregation and shape change
Catalog No. 50-297-3015 Supplier Cayman Chemical Supplier No. 148291 MG
An oxindole compound that potently blocks Syk activity (IC50 = 14 nM) and inhibits FcεRI-mediated rat RBL-2H3 basophil cell degranulation (EC50 = 313 nM); 2 μM completely abolishes convulxin-induced platelet aggregation and shape change