Learn More
Cayman Chemical Syk InhIbItr 500ug
SDP
Catalog No. 502971681
Encompass_Preferred
An oxindole compound that potently blocks Syk activity (IC50 = 14 nM) and inhibits FcεRI-mediated rat RBL-2H3 basophil cell degranulation (EC50 = 313 nM); 2 μM completely abolishes convulxin-induced platelet aggregation and shape change
Catalog No. 50-297-1681 Supplier Cayman Chemical Supplier No. 14829500 UG
An oxindole compound that potently blocks Syk activity (IC50 = 14 nM) and inhibits FcεRI-mediated rat RBL-2H3 basophil cell degranulation (EC50 = 313 nM); 2 μM completely abolishes convulxin-induced platelet aggregation and shape change