Learn More
Cayman Chemical Syk Inhibitor II, Purity: ≥98%, CAS: 726695-51-8, 5 mg
SDP
Catalog No. 502967413
Encompass_Preferred
A cell-permeable, selective, and reversible inhibitor of Syk (IC50 = 41 nM); prevents FcεRI-mediated 5-HT release in RBL-2H3 cells in vitro (IC50 = 460 nM) and inhibits passive cutaneous anaphylaxis reactions in mice (ID50 = 13.2 mg/kg, s.c.)
Catalog No. 50-296-7413 Supplier Cayman Chemical Supplier No. 182025 MG
A cell-permeable, selective, and reversible inhibitor of Syk (IC50 = 41 nM); prevents FcεRI-mediated 5-HT release in RBL-2H3 cells in vitro (IC50 = 460 nM) and inhibits passive cutaneous anaphylaxis reactions in mice (ID50 = 13.2 mg/kg, s.c.)