Learn More
Enzo Life Sciences Syk monoclonal antibody (SYK-01) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804480C100
Encompass_Preferred
Clone: SYK-01. Isotype: IgG1. Immunogen: Recombinant fragment (aa 5-360) of human Syk. Species reactivity: Human, Mouse, Rat. Applications: ICC, IHC (PS), IP, WB. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 15mM sodium azide.
Catalog No. 50-200-9920