Learn More
Bellco Glass, Inc. Sykes-Moore Chamber Only
SDP
Catalog No. 501944705
Encompass_Preferred
Study cells by high magnification, phase optics or time lapse. Cover-slips are separated by a silicone gasket to maintain internal seal. Sykes Moore wrench assembly is required and sold separately. 2.83 cm2 viewing area holds approximately 0.65 to 0.75mL. Made from brass. Chamber Only 0.250” x 1.250”
Catalog No. 50-194-4705 Supplier Bellco Glass, Inc. Supplier No. 194311111
Study cells by high magnification, phase optics or time lapse. Cover-slips are separated by a silicone gasket to maintain internal seal. Sykes Moore wrench assembly is required and sold separately. 2.83 cm2 viewing area holds approximately 0.65 to 0.75mL. Made from brass. Chamber Only 0.250” x 1.250”