Learn More
Bellco Glass, Inc. Sykes-Moore Gasket,1.5mm 1.5 mm Working Distance
SDP
Catalog No. 501944708
Encompass_Preferred
Sykes Moore gasket 1.5mm Dimensions: 1"x 7/8"x 1/16" thick Working distance 1.5mm in the Sykes Moore chamber
Catalog No. 50-194-4708 Supplier Bellco Glass, Inc. Supplier No. 194333315
Sykes Moore gasket 1.5mm Dimensions: 1"x 7/8"x 1/16" thick Working distance 1.5mm in the Sykes Moore chamber