Learn More
Bellco Glass, Inc. Sykes-Moore Holder For 1943-11111
SDP
Catalog No. 501944711
Encompass_Preferred
Ideal for viewing chamber on a microscope. Material: 2024-t4 alum Dimensions: 3" long x 1 1/2" wide x 3/16" thick Center hole size: 1.270" od x .094" dp
Catalog No. 50-194-4711 Supplier Bellco Glass, Inc. Supplier No. 194355555
Ideal for viewing chamber on a microscope. Material: 2024-t4 alum Dimensions: 3" long x 1 1/2" wide x 3/16" thick Center hole size: 1.270" od x .094" dp