Learn More
Bellco Glass, Inc. Sykes-Moore Wrench
SDP
Catalog No. 501944710
Encompass_Preferred
Ideal for holding the Sykes Moore culture chamber. Consist of two pieces, one flat aluminum holder, and a round aluminum chamber. Holder dimensions: 1 3/4" wide x 1 1/2" dp x 1/2" thick With notch: 1.270" x .875" (curved) Chamber holder: 1 1/4
Catalog No. 50-194-4710 Supplier Bellco Glass, Inc. Supplier No. 194344444
Ideal for holding the Sykes Moore culture chamber. Consist of two pieces, one flat aluminum holder, and a round aluminum chamber. Holder dimensions: 1 3/4" wide x 1 1/2" dp x 1/2" thick With notch: 1.270" x .875" (curved) Chamber holder: 1 1/4