Learn More
Ellsworth Adhesive Co Dow Sylgard 184 Silicone Encapsulant, Clear, 0.5 kg Kit
Catalog No. NC1508840
Encompass
The Dow Sylgard 184 Silicone Encapsulant Clear 0.5 kg Kit is a two-part silicone elastomer that cures at room temperature to a flexible elastomer. It is used for potting, encapsulating, coating, and mold making. The kit contains 0.5 kg of base and 0.5 kg of curing agent. It has a 1:1 mix ratio by weight, cures in 24 hours at 25C, has a viscosity of 3500 cP, tensile strength of 6.2 MPa, elongation of 170%, dielectric strength of 20 kV/mm, and volume resistivity of 2.9 x 1015 ohm cm. The operating temperature range is -45 to 200C. It has good adhesion and is resistant to moisture, dust, chemicals, and repeated thermal cycling.
Catalog No. NC1508840 Supplier Ellsworth Adhesive Co Supplier No. 40198620.5KG
The Dow Sylgard 184 Silicone Encapsulant Clear 0.5 kg Kit is a two-part silicone elastomer that cures at room temperature to a flexible elastomer. It is used for potting, encapsulating, coating, and mold making. The kit contains 0.5 kg of base and 0.5 kg of curing agent. It has a 1:1 mix ratio by weight, cures in 24 hours at 25C, has a viscosity of 3500 cP, tensile strength of 6.2 MPa, elongation of 170%, dielectric strength of 20 kV/mm, and volume resistivity of 2.9 x 1015 ohm cm. The operating temperature range is -45 to 200C. It has good adhesion and is resistant to moisture, dust, chemicals, and repeated thermal cycling.