Learn More
World Precision Instrument Sylgard Silicone Elastomer
SDP
Catalog No. 50822180
Encompass
 2-part silicone elastomer, ideal for potting and encapsulating applications
This silicone elastomer has a very low dielectric constant sealing compound, which makes it ideal for patch clamping and many other lab applications, especially in electrophysiology applications. 
Catalog No. 50-822-180 Supplier World Precision Instrument Supplier No. SYLG184
 2-part silicone elastomer, ideal for potting and encapsulating applications
This silicone elastomer has a very low dielectric constant sealing compound, which makes it ideal for patch clamping and many other lab applications, especially in electrophysiology applications. 