Learn More
RAVAGO CHEMICAL DISTRIBUTION INC SYLOID 161 Matting Agent, 50 lb bag
Catalog No. NC0576117
Encompass_Preferred
SYLOID 161 Matting Agent is an organically treated amorphous synthetic silica appearing as a white, free-flowing powder. This silica is a wax-treated matting, or gloss control agent designed for use in: general purpose OEM and DIY wood finishes, catalyzed varnishes (alkyd-urea, alkyd-amino, etc.), decorative enamels, both air-dry and forced-dry, DIY alkyd and urethane varnishes, general industrial coatings and UV curable coatings.
Catalog No. NC0576117 Supplier RAVAGO CHEMICAL DISTRIBUTION INC Supplier No. SYLOID161
SYLOID 161 Matting Agent is an organically treated amorphous synthetic silica appearing as a white, free-flowing powder. This silica is a wax-treated matting, or gloss control agent designed for use in: general purpose OEM and DIY wood finishes, catalyzed varnishes (alkyd-urea, alkyd-amino, etc.), decorative enamels, both air-dry and forced-dry, DIY alkyd and urethane varnishes, general industrial coatings and UV curable coatings.