Learn More
RAVAGO CHEMICAL DISTRIBUTION INC SYLOID 244, 30 lb bag
Catalog No. NC0410583
Encompass_Preferred
SYLOID 244 material is synthetic, amorphous silica appearing as a white free flowing powder. This product has a strong affinity for moisture and also for organic based liquids. SYLOID 244 silica can adsorb up to two and a half times its own weight in liquid. It is an amorphous silica ideal for application as a general purpose, high performance, anti-caking, free-flow and carrying agent for food and industrial applications.
Catalog No. NC0410583 Supplier RAVAGO CHEMICAL DISTRIBUTION INC Supplier No. SYLOID244
SYLOID 244 material is synthetic, amorphous silica appearing as a white free flowing powder. This product has a strong affinity for moisture and also for organic based liquids. SYLOID 244 silica can adsorb up to two and a half times its own weight in liquid. It is an amorphous silica ideal for application as a general purpose, high performance, anti-caking, free-flow and carrying agent for food and industrial applications.