Learn More
Loikits Distribution SYLTHERM 800-SILICONE LIQ 5GAL
Catalog No. NC9748497
Encompass
Fluid, Heat Transfer Fluid; Largeoikits; For Syltherm 800; Stable; Largeong lasting silicone based product; For high temp. liquid-phase operation; Temp: of - 40 deg. to 400 deg. C; High temp. stability; Largeo
Catalog No. NC9748497 Supplier Loikits Distribution Supplier No. SYLTHERM8005GAL
Fluid, Heat Transfer Fluid; Largeoikits; For Syltherm 800; Stable; Largeong lasting silicone based product; For high temp. liquid-phase operation; Temp: of - 40 deg. to 400 deg. C; High temp. stability; Largeo