Learn More
Chempoint Com Syltherm XLT, 5 gal
Catalog No. NC0504285
Encompass
Specifically formulated, high performance silicone polymer designed for use as a low temperature, liquid phase heat transfer medium. It offers remarkable low temperature heat transfer and pumpability, plus thermal stability.
Catalog No. NC0504285 Supplier Chempoint Com Supplier No. 0119591
Specifically formulated, high performance silicone polymer designed for use as a low temperature, liquid phase heat transfer medium. It offers remarkable low temperature heat transfer and pumpability, plus thermal stability.