Learn More
Bulbman Inc Osram 54189 64627HLX - EFP Bulb, 100 Watt, 12 Volt
SDP
Catalog No. 501253621
Encompass_Preferred
Osram EFP: GZ6.35 Base, MR16 Shape, 50 Hour Avg Lamp Life The Sylvania EFP 64627 HLX 100W/12V Lamp is a MR16 shape display optic halogen bulb. This lamp is suitable for 8mm projectors. Osram 54189 64627HLX - EFP bulb is a 100 watt 12 volt halogen specialty light bulb in the HLX Xenophot family of high performance Osram lamps. Osram HLX bulbs use xenon gas fill for increased luminous efficacy (up to 10% higher) and optimum color rendering for medical equipment and projector applications. The EFP bulb must be operated in a vertical (plus or minus 45 degrees) base down position. The bulb has a rated life of 50 hours.
Catalog No. 50-125-3621 Supplier Bulbman Inc Supplier No. 103233
Osram EFP: GZ6.35 Base, MR16 Shape, 50 Hour Avg Lamp Life The Sylvania EFP 64627 HLX 100W/12V Lamp is a MR16 shape display optic halogen bulb. This lamp is suitable for 8mm projectors. Osram 54189 64627HLX - EFP bulb is a 100 watt 12 volt halogen specialty light bulb in the HLX Xenophot family of high performance Osram lamps. Osram HLX bulbs use xenon gas fill for increased luminous efficacy (up to 10% higher) and optimum color rendering for medical equipment and projector applications. The EFP bulb must be operated in a vertical (plus or minus 45 degrees) base down position. The bulb has a rated life of 50 hours.