Learn More
Medchemexpress LLC SYM 2081 | 31137-74-3 | 98.0% | 161.16 g/mol | C6H11NO4 | 5 MG
SDP
Catalog No. 5000211256
Encompass_Preferred
SYM 2081 is a potent, selective kainate receptor agonist used as a research reagent to study excitatory neurotransmission, neuroprotection, and pain models. It modulates glutamate transport and elicits activity in kainate receptor assays; supplied as a purified solid for laboratory research (CAS 31137-74-3, formula C6H11NO4). For research use only.
  • Potent, selective agonist of kainate receptors.
  • Useful for studies of excitatory neurotransmission and synaptic physiology.
  • Supports research into neuroprotection and hypoxic-ischemic injury models.
  • High purity suitable for biochemical and pharmacological assays.
  • Supplied as a stable solid for storage and experimental use.
Catalog No. 50-002-11256 Supplier Medchemexpress LLC Supplier No. HY1013105MG
SYM 2081 is a potent, selective kainate receptor agonist used as a research reagent to study excitatory neurotransmission, neuroprotection, and pain models. It modulates glutamate transport and elicits activity in kainate receptor assays; supplied as a purified solid for laboratory research (CAS 31137-74-3, formula C6H11NO4). For research use only.
  • Potent, selective agonist of kainate receptors.
  • Useful for studies of excitatory neurotransmission and synaptic physiology.
  • Supports research into neuroprotection and hypoxic-ischemic injury models.
  • High purity suitable for biochemical and pharmacological assays.
  • Supplied as a stable solid for storage and experimental use.