Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Sym-Collidine, Reagent, pH 7.4 100 ML
SDP

Catalog No. 50980225
Encompass_Preferred

(2,4,6-Collidine; 2,4,6-Trimethylpyridine)
CAS #108-75-8 (C₅H₂N)(CH₃)₃ F.W. 121.18
Assay: 99%

Sym-Collidine is used in combination with osmium tetroxide to provide excellent fixation and stable buffering. Since the pH does not change when distilled water or osmium tetroxide solution is added, isotonic or hyper-tonic solutions can be made without recoursing the other solutes. Tissues treated with Sym-Collidine have an excellent, uniform consistency. Uranyl acetate stains very well and lead stain is not too intense.

The buffers are prepared fresh for each order and conveniently packaged. Desired pH should be specified when ordering.
pH: 7.4

Catalog No. 50-980-225 Supplier Electron Microscopy Sciences Supplier No. 11520
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2,4,6-Collidine; 2,4,6-Trimethylpyridine)
CAS #108-75-8 (C₅H₂N)(CH₃)₃ F.W. 121.18
Assay: 99%

Sym-Collidine is used in combination with osmium tetroxide to provide excellent fixation and stable buffering. Since the pH does not change when distilled water or osmium tetroxide solution is added, isotonic or hyper-tonic solutions can be made without recoursing the other solutes. Tissues treated with Sym-Collidine have an excellent, uniform consistency. Uranyl acetate stains very well and lead stain is not too intense.

The buffers are prepared fresh for each order and conveniently packaged. Desired pH should be specified when ordering.
pH: 7.4

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.