Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sym-Dicarbethoxyhydrazine 97%, Thermo Scientific™

Catalog No. AC113031000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC113031000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC113031000 Supplier Thermo Scientific Chemicals Supplier No. 113031000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4114-28-7
Molecular Formula C6H12N2O4
Molecular Weight (g/mol) 176.172
InChI Key JXMLAPZRDDWRRV-UHFFFAOYSA-N
Synonym diethyl 1,2-hydrazinedicarboxylate, diethyl hydrazodicarboxylate, 1,2-dicarbethoxyhydrazine, diethyl bicarbamate, 1,2-hydrazinedicarboxylic acid, diethyl ester, ethyl hydrazoformate, s-dicarbethoxyhydrazine, sym-dicarbethoxyhydrazine, bicarbamic acid, diethyl ester, n,n'-diethoxycarbonylhydrazine
PubChem CID 77758
IUPAC Name ethyl N-(ethoxycarbonylamino)carbamate
SMILES CCOC(=O)NNC(=O)OCC

Specifications

Melting Point 130°C to 135°C
Color White
Infrared Spectrum Authentic
Linear Formula C2H5O2CNHNHCO2C2H5
Quantity 100 g
Beilstein 03, 98
Formula Weight 176.17
Percent Purity ≥96.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material sym-Dicarbethoxyhydrazine, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.