Learn More
Medchemexpress LLC HY-18689 5mg Medchemexpress, SYM2206 CAS:173952-44-8 Purity:>98%
SDP
Catalog No. 502082072
Encompass_Preferred
Medchemexpress, HY-18689 5mg SYM2206 CAS:173952-44-8 SYM2206 is a potent and non-competitive AMPA receptor antagonist, with an IC50 of 1.6 μM. SYM2206 blocks Nav1.6-mediated persistent currents. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2072 Supplier Medchemexpress LLC Supplier No. HY186895MG
Medchemexpress, HY-18689 5mg SYM2206 CAS:173952-44-8 SYM2206 is a potent and non-competitive AMPA receptor antagonist, with an IC50 of 1.6 μM. SYM2206 blocks Nav1.6-mediated persistent currents. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.