Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Sym-Collidine Buffer, pH 7.4
SDP

Catalog No. 501898710
Encompass_Preferred

(2,4,6-Trimethylpyridine)
CAS #108-75-8 N:C(CH₃)CH:C(CH₃)CH:CCH₃ F.W. 121.18

A very stable buffer, used with Osmium tetroxide; provides excellent fixation, better stability and buffering capacity. The pH can be adjusted by varying the amount of hydrochloric acid in the final volume of 200 ml.
pH: 7.4

Catalog No. 50-189-8710 Supplier Electron Microscopy Sciences Supplier No. 11510
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2,4,6-Trimethylpyridine)
CAS #108-75-8 N:C(CH₃)CH:C(CH₃)CH:CCH₃ F.W. 121.18

A very stable buffer, used with Osmium tetroxide; provides excellent fixation, better stability and buffering capacity. The pH can be adjusted by varying the amount of hydrochloric acid in the final volume of 200 ml.
pH: 7.4

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.