Learn More
Waters Corp Symmetry Sentry Guard Cartridge, C18, 3.5 μm, 2.1 mm x 10 mm
Supplier:  Waters Corp WAT106127
Encompass_Preferred
This Symmetry C18 Sentry Guard Cartridge extends analytical column longevity by preventing suspended particles and chemical contaminants. Manufactured in a cGMP, ISO 9001-certified facility, it uses high-performance packing materials for consistent, reproducible results.
  • Features monofunctionally bonded C18 ligands on high purity base-deactivated silica.
  • Offers tight column specifications for highly reproducible chromatography.
  • Extends column longevity by preventing particles and contaminants.
  • Uses same high-performance packing as analytical columns.
  • Designed for greater capacity in biological fluid cleansing and/or temperature control.
  • Manufactured in a cGMP, ISO 9001-certified facility.
  • Packing material chromatographically tested per batch with neutral, acidic, and basic analytes.
  • Ensures consistently great, repeatable performance.
Catalog No. 50-819-045