Learn More
Waters Corp Symmetry C18 Sentry Guard Cartridge, 100Å, 5 µm, 3.9 mm X 20 mm, 2/pk
Catalog No. 50819044 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Symmetry C18 columns feature monofunctionally bonded C18 ligands on a high purity base-deactived silica. Symmetry columns have the tightest column specifications in the industry giving resulting highly reproducible chromatographic results.
Catalog No. 50-819-044 Supplier Waters Corp Supplier No. WAT054225
Symmetry C18 columns feature monofunctionally bonded C18 ligands on a high purity base-deactived silica. Symmetry columns have the tightest column specifications in the industry giving resulting highly reproducible chromatographic results.