Learn More
Waters Corp Symmetry C18 Column, 100 Å, 3.5 μm, 4.6 mm × 100 mm
Catalog No. 502391325 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Symmetry C18 Column is designed for high-performance liquid chromatography, featuring a 100 Å pore size and 3.5 μm particle size for effective separations. It comes with a 4.6 mm × 100 mm dimension and includes an eConnect Column Tag for enhanced traceability.
  • 100 Å pore size
  • 3.5 μm particle size
  • 4.6 mm × 100 mm dimensions
  • Supplied in a pack of one
  • Features an eConnect column tag
Catalog No. 50-239-1325 Supplier Waters Corp Supplier No. WAT066220RF
Add to Cart
Add to Cart
The Symmetry C18 Column is designed for high-performance liquid chromatography, featuring a 100 Å pore size and 3.5 μm particle size for effective separations. It comes with a 4.6 mm × 100 mm dimension and includes an eConnect Column Tag for enhanced traceability.
  • 100 Å pore size
  • 3.5 μm particle size
  • 4.6 mm × 100 mm dimensions
  • Supplied in a pack of one
  • Features an eConnect column tag