Learn More
Waters Corp HPLC Column, 100 Å, C18, 3.5 μm, 4.6 mm x 75 mm
Catalog No. 502391329 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This HPLC column is designed for high-performance liquid chromatography applications, featuring advanced eConnect technology for enhanced traceability and highly reproducible results due to its stringent manufacturing specifications.
  • Equipped with an electronic eConnect column tag that contains unique column-specific information.
  • NFC-enabled, device-tagged for automatic identification, tracking, and digital tracing of column use history.
  • Features trifunctionally bonded C18 ligands on a high-purity base-deactivated silica.
  • Known for having the tightest column specifications in the industry.
  • Provides highly reproducible chromatographic results.
Catalog No. 50-239-1329 Supplier Waters Corp Supplier No. WAT066224RF
This HPLC column is designed for high-performance liquid chromatography applications, featuring advanced eConnect technology for enhanced traceability and highly reproducible results due to its stringent manufacturing specifications.
  • Equipped with an electronic eConnect column tag that contains unique column-specific information.
  • NFC-enabled, device-tagged for automatic identification, tracking, and digital tracing of column use history.
  • Features trifunctionally bonded C18 ligands on a high-purity base-deactivated silica.
  • Known for having the tightest column specifications in the industry.
  • Provides highly reproducible chromatographic results.